Tisk…

4-(Cyanomethoxy)phenylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetonitrile

OR361030-1G OR361030-5G
APOSOR361030-1GEA 4370 CZK
APOSOR361030-1G APOSOR361030-5G
4-(Cyanomethoxy)phenylboronic acid pinacol ester
4-(Cyanomethoxy)phenylboronic acid pinacol ester
Vzorec: C₁₄H₁₈BNO₃
MW: 259.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795675
CAS číslo: 475272-13-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR