2,1,3-Benzoxadiazole-5-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Benzo[c][1,2,5]oxadiazole-5-carboxylic acid
OR3607-1G
OR3607-250MG
OR3607-5G
APOSOR3607-1GEA
744
CZK
APOSOR3607-1G
APOSOR3607-250MG
APOSOR3607-5G
2,1,3-Benzoxadiazole-5-carboxylic acid 97%
2,1,3-Benzoxadiazole-5-carboxylic acid
Vzorec:
C₇H₄N₂O₃ MW: 164,12 g/mol Bod tání: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00276977 CAS číslo: 19155-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...