Tisk…

Thieno[3,2-b]thiophene-2-boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: Thieno[3,2-b]thiophen-2-ylboronic acid

OR360558-1G OR360558-5G
APOSOR360558-1GEA 3110 CZK
APOSOR360558-1G APOSOR360558-5G
Thieno[3,2-b]thiophene-2-boronic acid
Thieno[3,2-b]thiophene-2-boronic acid
Vzorec: C₆H₅BO₂S₂
MW: 184,03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059468
CAS číslo: 160032-40-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR