Tisk…

4,4-Dimethylcyclohexene-1-ylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

OR360276-1G OR360276-5G
APOSOR360276-1GEA 1580 CZK
APOSOR360276-1G APOSOR360276-5G
4,4-Dimethylcyclohexene-1-ylboronic acid pinacol ester
4,4-Dimethylcyclohexene-1-ylboronic acid pinacol ester
Vzorec: C₁₄H₂₅BO₂
Storage Temperature: Freezer
MDL Number: MFCD08690233
CAS číslo: 859217-67-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR