Tisk…

2-Chloro-4-hydroxyphenylboronic acid

Dodavatel: Apollo Scientific

OR360269-1G OR360269-5G
APOSOR360269-1GEA 4040 CZK
APOSOR360269-1G APOSOR360269-5G
2-Chloro-4-hydroxyphenylboronic acid
2-Chloro-4-hydroxyphenylboronic acid
Vzorec: C₆H₆BClO₃
MW: 172,3740 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684320
CAS číslo: 766549-26-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR