Tisk…

3,4-Dichlorophenylboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific

OR360249-1G OR360249-5G
APOSOR360249-1GEA 1580 CZK
APOSOR360249-1G APOSOR360249-5G
3,4-Dichlorophenylboronic acid pinacol ester 98%
3,4-Dichlorophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₅BCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05663853
CAS číslo: 401797-02-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR