Tisk…

N-Methylpyrrole-3-boronic acid pinacol ester 97%

Dodavatel: Apollo Scientific

Synonyms: 1-METHYL-1H-PYRROL-3-YLBORONIC ACID PINACOL ESTER, 1H-Pyrrole,1-methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-, 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole

OR360175-1G
APOSOR360175-1GEA 1240 CZK
APOSOR360175-1G
N-Methylpyrrole-3-boronic acid pinacol ester 97%
N-Methylpyrrole-3-boronic acid pinacol ester
Vzorec: C₁₁H₁₈BNO₂
MW: 207,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182200
CAS číslo: 953040-54-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR