Tisk…

6-Bromo-2-pyridineboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR360154-10G OR360154-25G OR360154-5G
APOSOR360154-10GEA 3270 CZK
APOSOR360154-10G APOSOR360154-25G APOSOR360154-5G
6-Bromo-2-pyridineboronic acid pinacol ester 98%
6-Bromo-2-pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₅BBrNO₂
Bod tání: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD06796177
CAS číslo: 651358-83-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR