Tisk…

4-Quinolineboronic acid 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: Quinolin-4-ylboronic acid

OR360134-1G OR360134-5G
APOSOR360134-1GEA 1450 CZK
APOSOR360134-1G APOSOR360134-5G
4-Quinolineboronic acid 95+%
4-Quinolineboronic acid
Vzorec: C₉H₈BNO₂
MW: 172.9763 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03095174
CAS číslo: 371764-64-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR