Tisk…

3-(Trimethylsilyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-(Trimethylsilyl)phenyl)boronic acid

OR360037-1G OR360037-25G OR360037-5G
APOSOR360037-1GEA 622 CZK
APOSOR360037-1G APOSOR360037-25G APOSOR360037-5G
3-(Trimethylsilyl)phenylboronic acid 98%
3-(Trimethylsilyl)phenylboronic acid
Vzorec: C₉H₁₅BO₂Si
MW: 194,1100 g/mol
Bod tání: 145…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093884
CAS číslo: 177171-16-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR