5-Oxazolecarboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Oxazole-5-carboxylic acid
OR3589-500MG
OR3589-25G
OR3589-5G
APOSOR3589-500MGEA
1040
CZK
APOSOR3589-500MG
APOSOR3589-25G
APOSOR3589-5G
5-Oxazolecarboxylic acid 98%
5-Oxazolecarboxylic acid
Vzorec:
C₄H₃NO₃ MW: 113.07 g/mol Bod tání: 226…230 °C Storage Temperature: Freezer |
MDL Number:
MFCD04114931 CAS číslo: 118994-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...