Methyl-5-(ethylsulphonyl)-2-methoxybenzoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-(Ethylsulphonyl)-2-methoxybenzoic Acid Methyl Ester
OR3571-100G
OR3571-25G
OR3571-5G
APOSOR3571-100GEA
9760
CZK
APOSOR3571-100G
APOSOR3571-25G
APOSOR3571-5G
Methyl-5-(ethylsulphonyl)-2-methoxybenzoate
Methyl-5-(ethylsulphonyl)-2-methoxybenzoate
Vzorec:
C₁₁H₁₄O₅S MW: 258.29 g/mol Bod tání: 126 °C Storage Temperature: Ambient |
MDL Number:
MFCD01317547 CAS číslo: 62140-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...