2-Methoxy-5-sulphamoylbenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR3567-10G
OR3567-50G
OR3567-100G
APOSOR3567-10GEA
662
CZK
APOSOR3567-10G
APOSOR3567-50G
APOSOR3567-100G
2-Methoxy-5-sulphamoylbenzoic acid
2-Methoxy-5-sulphamoylbenzoic acid
Vzorec:
C₈H₉NO₅S MW: 232,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00129997 CAS číslo: 22117-85-7 EINECS: 244-789-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...