Tisk…

Methyl-3-(chlorosulphonyl)benzoate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 3-(chlorosulphonyl)benzoate

OR3564-1G OR3564-25G OR3564-5G
APOSOR3564-1GEA 870 CZK
APOSOR3564-1G APOSOR3564-25G APOSOR3564-5G
Methyl-3-(chlorosulphonyl)benzoate
Methyl-3-(chlorosulphonyl)benzoate
Vzorec: C₈H₇ClO₄S
Bod tání: 63…69 °C
Storage Temperature: Ambient
MDL Number: MFCD06408800
CAS číslo: 63555-50-0
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR