2-Acetamidothiazole 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR3555-25G
OR3555-100G
APOSOR3555-25GEA
1820
CZK
APOSOR3555-25G
APOSOR3555-100G
2-Acetamidothiazole 96%
2-Acetamidothiazole
Vzorec:
C₅H₆N₂OS MW: 142.18 g/mol Bod tání: 204…206 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016892 CAS číslo: 2719-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...