Tisk…

6-Iodonicotinonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR3547-1G OR3547-5G
APOSOR3547-1GEA 5860 CZK
APOSOR3547-1G APOSOR3547-5G
6-Iodonicotinonitrile 98%
6-Iodonicotinonitrile
Vzorec: C₆H₃IN₂
MW: 230.0100 g/mol
Bod tání: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD08689512
CAS číslo: 289470-22-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR