Tisk…

2-Bromo-4'-(dimethylamino)acetophenone 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Dimethylamino)phenacyl bromide, 2-Bromo-1-(4-(dimethylamino)phenyl)ethanone

OR3530-1G OR3530-250MG OR3530-5G
APOSOR3530-1GEA 1620 CZK
APOSOR3530-1G APOSOR3530-250MG APOSOR3530-5G
2-Bromo-4'-(dimethylamino)acetophenone 95%
2-Bromo-4'-(dimethylamino)acetophenone
Vzorec: C₁₀H₁₂BrNO
MW: 242,1124 g/mol
Bod tání: 89…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409221
CAS číslo: 37904-72-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR