Tisk…

6-Bromo-7-methylimidazo[1,2-a]pyridine 98%

Dodavatel: Apollo Scientific
Varování

OR3506-1G OR3506-5G OR3506-25G
APOSOR3506-1GEA 1410 CZK
APOSOR3506-1G APOSOR3506-5G APOSOR3506-25G
6-Bromo-7-methylimidazo[1,2-a]pyridine 98%
6-Bromo-7-methylimidazo[1,2-a]pyridine
Vzorec: C₈H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09258769
CAS číslo: 116355-18-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR