N-Tosyl-pyrrole-3-carboxylic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR350522-1G
OR350522-5G
APOSOR350522-1GEA
744
CZK
APOSOR350522-1G
APOSOR350522-5G
N-Tosyl-pyrrole-3-carboxylic acid 96%
N-Tosyl-pyrrole-3-carboxylic acid
Vzorec:
C₁₂H₁₁NO₄S MW: 265,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00145013 CAS číslo: 106058-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...