4-Chloroquinoline-6-carbonitrile
Dodavatel: Apollo Scientific
OR350463-1G
OR350463-250MG
APOSOR350463-1GEA
11580
CZK
APOSOR350463-1G
APOSOR350463-250MG
4-Chloroquinoline-6-carbonitrile
4-Chloroquinoline-6-carbonitrile
Vzorec:
C₁₀H₅ClN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08063204 CAS číslo: 219763-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...