Tisk…

2-(Quinolin-6-yl)acetic acid

Dodavatel: Apollo Scientific

OR350332-1G OR350332-5G
APOSOR350332-1GEA 655 CZK
APOSOR350332-1G APOSOR350332-5G
2-(Quinolin-6-yl)acetic acid
2-(Quinolin-6-yl)acetic acid
Vzorec: C₁₁H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD07776794
CAS číslo: 5622-34-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR