Tisk…

Pyridinium-p-toluenesulphonate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Pyridin-1-ium 4-methylbenzenesulphonate

OR350313-500G
APOSOR350313-500GEA 2280 CZK
APOSOR350313-500G
Pyridinium-p-toluenesulphonate 98%
Pyridinium-p-toluenesulphonate
Vzorec: C₁₂H₁₃NO₃S
MW: 251,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013108
CAS číslo: 24057-28-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR