(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 97%
Dodavatel: Apollo Scientific
Synonyms:
H-Lys(Z)-OBzl.HCl, H-Lys(Z)-OBzl·HCl, Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride
OR350309-5G
OR350309-1G
OR350309-25G
APOSOR350309-5GEA
2900
CZK
APOSOR350309-5G
APOSOR350309-1G
APOSOR350309-25G
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 97%
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Vzorec:
C₂₁H₂₆N₂O₄·HCl MW: 406,9 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038981 CAS číslo: 6366-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...