5-Methoxynicotinonitrile 98%
Dodavatel: Apollo Scientific
OR350274-5G
APOSOR350274-5GEA
6550
CZK
APOSOR350274-5G
5-Methoxynicotinonitrile 98%
5-Methoxynicotinonitrile
Vzorec:
C₇H₆N₂O Storage Temperature: Ambient |
MDL Number:
MFCD09909444 CAS číslo: 298204-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...