Tisk…

Methyl-1,2,4-triazole-3-carboxylate 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate

OR350262-500G
APOSOR350262-500GEA 2110 CZK
APOSOR350262-500G
Methyl-1,2,4-triazole-3-carboxylate 97%
Methyl-1,2,4-triazole-3-carboxylate
Vzorec: C₄H₅N₃O₂
MW: 127,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135989
CAS číslo: 4928-88-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR