Tisk…

4-(Methylamino)-3-nitrobenzoic acid 98%

Dodavatel: Apollo Scientific
Varování

OR350255-100G OR350255-25G
APOSOR350255-100GEA 2240 CZK
APOSOR350255-100G APOSOR350255-25G
4-(Methylamino)-3-nitrobenzoic acid 98%
4-(Methylamino)-3-nitrobenzoic acid
Vzorec: C₈H₈N₂O₄
MW: 196,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464322
CAS číslo: 41263-74-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR