Tisk…

3-Methyl-6-nitro-1H-indazole 99%

Dodavatel: Apollo Scientific
Varování

OR350252-100G OR350252-25G OR350252-5G
APOSOR350252-100GEA 2570 CZK
APOSOR350252-100G APOSOR350252-25G APOSOR350252-5G
3-Methyl-6-nitro-1H-indazole 99%
3-Methyl-6-nitro-1H-indazole
Vzorec: C₈H₇N₃O₂
MW: 177,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07082704
CAS číslo: 6494-19-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR