α-Bromo-o-xylene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Methylbenzyl bromide, alpha-Bromo-o-xylene
OR350233-100G
OR350233-25G
APOSOR350233-100GEA
13230
CZK
APOSOR350233-100G
APOSOR350233-25G
α-Bromo-o-xylene 98%
α-Bromo-o-xylene
Vzorec:
C₈H₉Br MW: 185.06 Bod varu: 216…217 °C Bod tání: 16…20 °C Hustota: 1.381 g/cm³ (25 °C) Bod vzplanutí: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000174 CAS číslo: 89-92-9 EINECS: 201-951-6 UN: 1701 ADR: 6.1 II Merck Index: 12,10223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...