Tisk…

3-Iodo-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Iodopyrazole

OR350207-25G OR350207-5G OR350207-10G
APOSOR350207-25GEA 4270 CZK
APOSOR350207-25G APOSOR350207-5G APOSOR350207-10G
3-Iodo-1H-pyrazole
3-Iodo-1H-pyrazole
Vzorec: C₃H₃IN₂
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS číslo: 4522-35-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR