4-(Hydroxymethyl)benzoic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
4-Carboxybenzylalcohol, HMBA Linker, HMBA
OR350195-25G
OR350195-5G
OR350195-100G
APOSOR350195-25GEA
2150
CZK
APOSOR350195-25G
APOSOR350195-5G
APOSOR350195-100G
4-(Hydroxymethyl)benzoic acid 98%
4-(Hydroxymethyl)benzoic acid
Vzorec:
C₈H₈O₃ MW: 152,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017598 CAS číslo: 3006-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...