Tisk…

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine 97%

Dodavatel: Apollo Scientific

Synonyms: Fmoc-Thr(tBu)-OH, Nalpha-Fmoc-O-tert-butyl-L-threonine, O-tert-Butyl-Nalpha-Fmoc-L-threonine, FMOC-O-tert-Butyl-L-threonine

OR350183-100G OR350183-500G
APOSOR350183-100GEA 2650 CZK
APOSOR350183-100G APOSOR350183-500G
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine 97%
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine
Vzorec: C₂₃H₂₇NO₅
MW: 397.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077075
CAS číslo: 71989-35-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR