Tisk…

2-Ethylhexyl-2-cyano-3,3-diphenylacrylate

Dodavatel: Apollo Scientific

Synonyms: 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, Octyl 2-Cyano-3,3-diphenylacrylate, 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octocrylene

OR350157-100G OR350157-5G OR350157-25G
APOSOR350157-100GEA 1870 CZK
APOSOR350157-100G APOSOR350157-5G APOSOR350157-25G
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate
Vzorec: C₂₄H₂₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00059260
CAS číslo: 6197-30-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR