Tisk…

6-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde

Dodavatel: Apollo Scientific
Varování

OR350033-1G OR350033-5G
APOSOR350033-1GEA 2980 CZK
APOSOR350033-1G APOSOR350033-5G
6-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde
6-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde
Vzorec: C₈H₅BrN₂O
MW: 225,0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09864909
CAS číslo: 30384-96-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR