tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
[1-(Hydroxymethyl)cyclopropyl]carbamic acid tert-Butyl ester, [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(tert-Butoxycarbonylamino)cyclopropyl]methanol
OR350017-1G
OR350017-5G
OR350017-10G
APOSOR350017-1GEA
622
CZK
APOSOR350017-1G
APOSOR350017-5G
APOSOR350017-10G
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate 98%
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate
Vzorec:
C₉H₁₇NO₃ MW: 187.2362 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09749954 CAS číslo: 107017-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...