Tisk…
1,3-Adamantanedicarboxylic acid 97%
Dodavatel: Apollo Scientific
Synonyms: 1,3-Adamantanedicarboxylic Acid, 1,3-Dicarboxyadamantane, (1s,3s,5r,7r)-Adamantane-1,3-dicarboxylic acid
OR350008-25G OR350008-100G
APOSOR350008-25GEA 2530 CZK
APOSOR350008-25G APOSOR350008-100G
1,3-Adamantanedicarboxylic acid 97%
1,3-Adamantanedicarboxylic acid
Vzorec: C₁₂H₁₆O₄
MW: 224.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167790
CAS číslo: 39269-10-8