Tisk…

2-Methoxy-4-pyridineboronic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Methoxy-4-pyridylboronic acid, 2-Methoxypyridine-4-boronic acid, (2-Methoxypyridin-4-yl)boronic acid

OR3487-10G OR3487-1G OR3487-5G
APOSOR3487-10GEA 5590 CZK
APOSOR3487-10G APOSOR3487-1G APOSOR3487-5G
2-Methoxy-4-pyridineboronic acid 97%
2-Methoxy-4-pyridineboronic acid
Vzorec: C₆H₈BNO₃
MW: 152,9436 g/mol
Bod tání: 65…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368877
CAS číslo: 762262-09-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR