Tisk…

5-Bromo-2-methoxy-3-pyridineboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-methoxy-3-pyridinylboronic acid, 5-Bromo-2-methoxypyridine-3-boronic acid

OR3483-1G OR3483-5G OR3483-25G
APOSOR3483-1GEA 5030 CZK
APOSOR3483-1G APOSOR3483-5G APOSOR3483-25G
5-Bromo-2-methoxy-3-pyridineboronic acid 97%
5-Bromo-2-methoxy-3-pyridineboronic acid
Vzorec: C₆H₇BBrNO₃
MW: 231,84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973352
CAS číslo: 850864-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR