Ethyl-4-amino-3-chlorobenzoate 95+%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 4-amino-3-chlorobenzoate, 4-Amino-3-chlorobenzoic acid ethyl ester
OR346587-5G
APOSOR346587-5GEA
1660
CZK
APOSOR346587-5G
Ethyl-4-amino-3-chlorobenzoate 95+%
Ethyl-4-amino-3-chlorobenzoate
Vzorec:
C₉H₁₀ClNO₂ MW: 199,6300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06411007 CAS číslo: 82765-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...