4-Bromo-3-methylisothiazole 95+%
Dodavatel: Apollo Scientific
OR346459-1G
OR346459-500MG
APOSOR346459-1GEA
27910
CZK
APOSOR346459-1G
APOSOR346459-500MG
4-Bromo-3-methylisothiazole 95+%
4-Bromo-3-methylisothiazole
Vzorec:
C₄H₄BrNS Storage Temperature: Ambient |
MDL Number:
MFCD11110311 CAS číslo: 930-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...