1-p-Tolyl-1H-pyrazole-4-carbaldehyde 95+%
Dodavatel: Apollo Scientific
Synonyms:
1-(p-Tolyl)-1H-pyrazole-4-carbaldehyde
OR346440-1G
OR346440-500MG
OR346440-5G
APOSOR346440-1GEA
8370
CZK
APOSOR346440-1G
APOSOR346440-500MG
APOSOR346440-5G
1-p-Tolyl-1H-pyrazole-4-carbaldehyde 95+%
1-p-Tolyl-1H-pyrazole-4-carbaldehyde
Vzorec:
C₁₁H₁₀N₂O MW: 186,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426978 CAS číslo: 337957-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...