Tisk…

4-tert-Butylbenzamidoxime

Dodavatel: Apollo Scientific

Synonyms: 4-tert-Butyl-N'-hydroxy-benzamidine

OR346324-1G OR346324-5G
APOSOR346324-1GEA 0 CZK
APOSOR346324-1G APOSOR346324-5G
4-tert-Butylbenzamidoxime
4-tert-Butylbenzamidoxime
Vzorec: C₁₁H₁₆N₂O
Storage Temperature: Ambient
CAS číslo: 175204-39-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR