Tisk…

1-Quinoxalin-2-ylbutane-1,2,3,4-tetraol

Dodavatel: Apollo Scientific
Varování

OR346271-1G OR346271-5G
APOSOR346271-1GEA 0 CZK
APOSOR346271-1G APOSOR346271-5G
1-Quinoxalin-2-ylbutane-1,2,3,4-tetraol
1-Quinoxalin-2-ylbutane-1,2,3,4-tetraol
Vzorec: C₁₂H₁₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD01096782
CAS číslo: 80840-09-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR