3-Phenyl-1,2,4-oxadiazol-5-ol
Dodavatel: Apollo Scientific
OR346246-1G
OR346246-5G
APOSOR346246-1GEA
2730
CZK
APOSOR346246-1G
APOSOR346246-5G
3-Phenyl-1,2,4-oxadiazol-5-ol
3-Phenyl-1,2,4-oxadiazol-5-ol
Vzorec:
C₈H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD17676121 CAS číslo: 1456-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...