2-Chloro-4-methylpyridine-3-sulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR346211-1G
OR346211-500MG
APOSOR346211-1GEA
6220
CZK
APOSOR346211-1G
APOSOR346211-500MG
2-Chloro-4-methylpyridine-3-sulphonyl chloride
2-Chloro-4-methylpyridine-3-sulphonyl chloride
Vzorec:
C₆H₅Cl₂NO₂S MW: 226.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15142814 CAS číslo: 1208081-91-7 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...