2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole
Dodavatel: Apollo Scientific
OR346176-1G
OR346176-500MG
OR346176-5G
APOSOR346176-1GEA
4530
CZK
APOSOR346176-1G
APOSOR346176-500MG
APOSOR346176-5G
2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole
2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole
Vzorec:
C₉H₉ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00457523 CAS číslo: 4760-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...