4-(5-Chloro[1,2,4]thiadiazol-3-yl)morpholine
Dodavatel: Apollo Scientific
OR346175-1G
OR346175-500MG
APOSOR346175-1GEA
0
CZK
APOSOR346175-1G
APOSOR346175-500MG
4-(5-Chloro[1,2,4]thiadiazol-3-yl)morpholine
4-(5-Chloro[1,2,4]thiadiazol-3-yl)morpholine
Vzorec:
C₆H₈ClN₃OS MW: 205,67 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12131114 CAS číslo: 887623-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...