2-(5-Chloro[1,2,4]thiadiazol-3-yl)pyridine
Dodavatel: Apollo Scientific
OR346174-250MG
OR346174-500MG
APOSOR346174-250MGEA
0
CZK
APOSOR346174-250MG
APOSOR346174-500MG
2-(5-Chloro[1,2,4]thiadiazol-3-yl)pyridine
2-(5-Chloro[1,2,4]thiadiazol-3-yl)pyridine
Vzorec:
C₇H₄ClN₃S Storage Temperature: Ambient |
MDL Number:
MFCD12131112 CAS číslo: 887623-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...