2,2-Dibromo-3-oxobutyric acid ethyl ester
Dodavatel: Apollo Scientific
OR346159-1G
OR346159-5G
APOSOR346159-1GEA
0
CZK
APOSOR346159-1G
APOSOR346159-5G
2,2-Dibromo-3-oxobutyric acid ethyl ester
2,2-Dibromo-3-oxobutyric acid ethyl ester
Vzorec:
C₆H₈Br₂O₃ MW: 287,95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11845706 CAS číslo: 89415-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...