Tisk…

5-Nitrobenzo[d]thiazole

Dodavatel: Apollo Scientific

OR346149-1G
APOSOR346149-1GEA 6910 CZK
APOSOR346149-1G
5-Nitrobenzo[d]thiazole
5-Nitrobenzo[d]thiazole
Vzorec: C7H4N2O2S
MW: 180,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085891
CAS číslo: 2942-07-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR