5-Chloro-2-cyclopropyl-6-methylpyrimidin-4-ol
Dodavatel: Apollo Scientific
OR346125-1G
OR346125-5G
APOSOR346125-1GEA
0
CZK
APOSOR346125-1G
APOSOR346125-5G
5-Chloro-2-cyclopropyl-6-methylpyrimidin-4-ol
5-Chloro-2-cyclopropyl-6-methylpyrimidin-4-ol
Vzorec:
C₈H₉ClN₂O MW: 184,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00114339 CAS číslo: 287927-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...